Anti-LCN2 Rat Monoclonal Antibody (Biotin) [clone: 228418]

Supplier: R & D Systems
BAM1857
102879-118EA 746.22 USD
102879-118
Anti-LCN2 Rat Monoclonal Antibody (Biotin) [clone: 228418]
Antibodies
Type: Primary
Antigen: LCN2
Clonality: Monoclonal
Clone: 228418
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR