Anti-IL6RA Rat Monoclonal Antibody (Biotin) [clone: 255821]

Supplier: R & D Systems
BAM1830
220014-582EA 698.49 USD
220014-582
Anti-IL6RA Rat Monoclonal Antibody (Biotin) [clone: 255821]
Antibodies
Type: Primary
Antigen: IL6RA
Clonality: Monoclonal
Clone: 255821
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR