Anti-IL5 Mouse Monoclonal Antibody (Biotin) [clone: 604308]

Supplier: R & D Systems
BAM17951
102879-116EA 746.22 USD
102879-116
Anti-IL5 Mouse Monoclonal Antibody (Biotin) [clone: 604308]
Antibodies
Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone: 604308
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR