Anti-IL28A/B Rat Monoclonal Antibody (Biotin) [clone: 244707]

Supplier: R & D Systems
BAM17891
220014-581EA 746.22 USD
220014-581
Anti-IL28A/B Rat Monoclonal Antibody (Biotin) [clone: 244707]
Antibodies
Type: Primary
Antigen: IL28A/B
Clonality: Monoclonal
Clone: 244707
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR