Anti-DKK1 Rat Monoclonal Antibody (Biotin) [clone: 259631]

Supplier: R & D Systems
BAM1765
220014-580EA 689.82 USD
220014-580
Anti-DKK1 Rat Monoclonal Antibody (Biotin) [clone: 259631]
Antibodies
Type: Primary
Antigen: DKK1
Clonality: Monoclonal
Clone: 259631
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR