Anti-CRP Mouse Monoclonal Antibody (Biotin) [clone: 233520]

Supplier: R & D Systems
BAM17442
220014-579EA 746.22 USD
220014-579
Anti-CRP Mouse Monoclonal Antibody (Biotin) [clone: 233520]
Antibodies
Type: Primary
Antigen: CRP
Clonality: Monoclonal
Clone: 233520
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR