Anti-FETUB Mouse Monoclonal Antibody (Biotin) [clone: 212633]

Supplier: R & D Systems
BAM17251
220014-578EA 728.86 USD
220014-578
Anti-FETUB Mouse Monoclonal Antibody (Biotin) [clone: 212633]
Antibodies
Type: Primary
Antigen Symbol: FETUB
Antigen Name: fetuin B
Antigen Synonyms: 16G2, Gugu, IRL685
Clonality: Monoclonal
Clone: 212633
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR