Anti-pY Mouse Monoclonal Antibody (Biotin) [clone: 179003]

Supplier: R & D Systems
BAM1676
220014-575EA 451.2 USD
220014-575
Anti-pY Mouse Monoclonal Antibody (Biotin) [clone: 179003]
Antibodies
Type: Primary
Antigen: pY
Clonality: Monoclonal
Clone: 179003
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR