Anti-CXCL8 Mouse Monoclonal Antibody (Biotin) [clone: 258911]

Supplier: R & D Systems
BAM16082
220014-574EA 714.4 USD
220014-574
Anti-CXCL8 Mouse Monoclonal Antibody (Biotin) [clone: 258911]
Antibodies
Type: Primary
Antigen: CXCL8
Clonality: Monoclonal
Clone: 258911
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR