Anti-CD163 Mouse Monoclonal Antibody (Biotin) [clone: 215901]

Supplier: R & D Systems
BAM16072
220014-573EA 762.12 USD
220014-573
Anti-CD163 Mouse Monoclonal Antibody (Biotin) [clone: 215901]
Antibodies
Type: Primary
Antigen: CD163
Clonality: Monoclonal
Clone: 215901
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR