Anti-AHSG Rat Monoclonal Antibody (Biotin) [clone: 27432]

Supplier: R & D Systems
BAM15631
220014-572EA 746.22 USD
220014-572
Anti-AHSG Rat Monoclonal Antibody (Biotin) [clone: 27432]
Antibodies
Type: Primary
Antigen: AHSG
Clonality: Monoclonal
Clone: 27432
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR