Anti-KLRK1 Rat Monoclonal Antibody (Biotin) [clone: 191004]

Supplier: R & D Systems
BAM1547
220014-571EA 714.4 USD
220014-571
Anti-KLRK1 Rat Monoclonal Antibody (Biotin) [clone: 191004]
Antibodies
Type: Primary
Antigen: KLRK1
Clonality: Monoclonal
Clone: 191004
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR