Anti-INSR Mouse Monoclonal Antibody (Biotin) [clone: 243524]

Supplier: R & D Systems
BAM1544
220014-570EA 561.11 USD
220014-570
Anti-INSR Mouse Monoclonal Antibody (Biotin) [clone: 243524]
Antibodies
Type: Primary
Antigen: INSR
Clonality: Monoclonal
Clone: 243524
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR