Anti-ALPL Mouse Monoclonal Antibody (Biotin) [clone: B4-78]

Supplier: R & D Systems
BAM1448
220014-569EA 561.11 USD
220014-569
Anti-ALPL Mouse Monoclonal Antibody (Biotin) [clone: B4-78]
Antibodies
Type: Primary
Antigen: ALPL
Clonality: Monoclonal
Clone: B4-78
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR