Anti-SSEA4 Mouse Monoclonal Antibody (Biotin) [clone: MC-813-70]

Supplier: R & D Systems
BAM1435
220014-567EA 594.37 USD
220014-567
Anti-SSEA4 Mouse Monoclonal Antibody (Biotin) [clone: MC-813-70]
Antibodies
Type: Primary
Antigen: SSEA4
Clonality: Monoclonal
Clone: MC-813-70
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR