Anti-DCN Mouse Monoclonal Antibody (Biotin) [clone: 115413]

Supplier: R & D Systems
BAM1431
220014-566EA 740.43 USD
220014-566
Anti-DCN Mouse Monoclonal Antibody (Biotin) [clone: 115413]
Antibodies
Type: Primary
Antigen: DCN
Clonality: Monoclonal
Clone: 115413
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR