Anti-SOST Mouse Monoclonal Antibody (Biotin) [clone: 220910]

Supplier: R & D Systems
BAM14061
102879-114EA 746.22 USD
102879-114
Anti-SOST Mouse Monoclonal Antibody (Biotin) [clone: 220910]
Antibodies
Type: Primary
Antigen: SOST
Clonality: Monoclonal
Clone: 220910
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR