Anti-CD80 Mouse Monoclonal Antibody (Biotin) [clone: 37721]

Supplier: R & D Systems
BAM1402
220014-562EA 698.49 USD
220014-562
Anti-CD80 Mouse Monoclonal Antibody (Biotin) [clone: 37721]
Antibodies
Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 37721
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR