Anti-DLL4 Rat Monoclonal Antibody (Biotin) [clone: 207803]

Supplier: R & D Systems
BAM13892
220014-561EA 746.22 USD
220014-561
Anti-DLL4 Rat Monoclonal Antibody (Biotin) [clone: 207803]
Antibodies
Type: Primary
Antigen: DLL4
Clonality: Monoclonal
Clone: 207803
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR