Anti-RETN Mouse Monoclonal Antibody (Biotin) [clone: 184320]

Supplier: R & D Systems
BAM1359
220014-560EA 614.62 USD
220014-560
Anti-RETN Mouse Monoclonal Antibody (Biotin) [clone: 184320]
Antibodies
Type: Primary
Antigen: RETN
Clonality: Monoclonal
Clone: 184320
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR