Anti-INS Mouse Monoclonal Antibody (Biotin) [clone: 253627]

Supplier: R & D Systems
BAM13361
220014-554EA 543.75 USD
220014-554
Anti-INS Mouse Monoclonal Antibody (Biotin) [clone: 253627]
Antibodies
Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: 253627
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR