Anti-JAG1 Mouse Monoclonal Antibody (Biotin) [clone: 188331]

Supplier: R & D Systems
BAM1277
220014-553EA 681.14 USD
220014-553
Anti-JAG1 Mouse Monoclonal Antibody (Biotin) [clone: 188331]
Antibodies
Type: Primary
Antigen: JAG1
Clonality: Monoclonal
Clone: 188331
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR