Anti-LGALS1 Rat Monoclonal Antibody (Biotin) [clone: 201002]

Supplier: R & D Systems
BAM12451
220014-549EA 740.43 USD
220014-549
Anti-LGALS1 Rat Monoclonal Antibody (Biotin) [clone: 201002]
Antibodies
Type: Primary
Antigen: LGALS1
Clonality: Monoclonal
Clone: 201002
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR