Anti-TNFSF4 Rat Monoclonal Antibody (Biotin) [clone: 182603]

Supplier: R & D Systems
BAM12361
220014-548EA 746.22 USD
220014-548
Anti-TNFSF4 Rat Monoclonal Antibody (Biotin) [clone: 182603]
Antibodies
Type: Primary
Antigen: TNFSF4
Clonality: Monoclonal
Clone: 182603
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR