Anti-GYPA Mouse Monoclonal Antibody (Biotin) [clone: R10]

Supplier: R & D Systems
BAM12281
220014-546EA 740.43 USD
220014-546
Anti-GYPA Mouse Monoclonal Antibody (Biotin) [clone: R10]
Antibodies
Type: Primary
Antigen Symbol: GYPA
Antigen Name: glycophorin A (MNS blood group)
Antigen Synonyms: CD235a, GPA, GPErik, GPSAT, HGpMiV, HGpMiXI, HGpSta(C), MN, MNS, PAS-2
Clonality: Monoclonal
Clone: R10
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR