Anti-CST3 Mouse Monoclonal Antibody (Biotin) [clone: 197821]

Supplier: R & D Systems
BAM11961
220014-542EA 740.43 USD
220014-542
Anti-CST3 Mouse Monoclonal Antibody (Biotin) [clone: 197821]
Antibodies
Type: Primary
Antigen: CST3
Clonality: Monoclonal
Clone: 197821
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR