Anti-Neuron-specific beta-III Tubulin Mouse Monoclonal Antibody [clone: TuJ-1] (Biotin)

Supplier: R & D Systems
BAM1195
220014-541EA 723.08 USD
220014-541
Anti-Neuron-specific beta-III Tubulin Mouse Monoclonal Antibody [clone: TuJ-1] (Biotin)
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: Neuron-specific beta-III Tubulin
Antigen Synonyms: Tubulin Beta 3, Tubulin beta-4
Clonality: Monoclonal
Clone: TuJ-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR