Anti-AGER Rat Monoclonal Antibody (Biotin) [clone: 17544]

Supplier: R & D Systems
BAM11791
220014-539EA 714.4 USD
220014-539
Anti-AGER Rat Monoclonal Antibody (Biotin) [clone: 17544]
Antibodies
Type: Primary
Antigen: AGER
Clonality: Monoclonal
Clone: 17544
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR