Anti-CD8A Rat Monoclonal Antibody (Biotin) [clone: 53-6.7]

Supplier: R & D Systems
BAM116
220014-538EA 728.86 USD
220014-538
Anti-CD8A Rat Monoclonal Antibody (Biotin) [clone: 53-6.7]
Antibodies
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR