Anti-CD5 Rat Monoclonal Antibody (Biotin) [clone: 53-7.3]

Supplier: R & D Systems
BAM115
220014-537EA 728.86 USD
220014-537
Anti-CD5 Rat Monoclonal Antibody (Biotin) [clone: 53-7.3]
Antibodies
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 53-7.3
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR