Anti-PTPRC Rat Monoclonal Antibody (Biotin) [clone: 30-F11]

Supplier: R & D Systems
BAM114
220014-536EA 728.86 USD
220014-536
Anti-PTPRC Rat Monoclonal Antibody (Biotin) [clone: 30-F11]
Antibodies
Complement-dependent Cytotoxicity,

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 30-F11
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR