Anti-ITGAM Rat Monoclonal Antibody (Biotin) [clone: M1/70]

Supplier: R & D Systems
BAM1124
220014-534EA 728.86 USD
220014-534
Anti-ITGAM Rat Monoclonal Antibody (Biotin) [clone: M1/70]
Antibodies
Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: M1/70
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR