Anti-FZD1 Rat Monoclonal Antibody (Biotin) [clone: 162531]

Supplier: R & D Systems
BAM11201
220014-533EA 543.75 USD
220014-533
Anti-FZD1 Rat Monoclonal Antibody (Biotin) [clone: 162531]
Antibodies
Type: Primary
Antigen Symbol: FZD1
Antigen Name: frizzled homolog 1 (Drosophila)
Antigen Synonyms: AW227548, FZ-1, Fz1
Clonality: Monoclonal
Clone: 162531
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR