Anti-ENG Mouse Monoclonal Antibody (Biotin) [clone: 166707]

Supplier: R & D Systems
BAM10971
220014-532EA 594.37 USD
220014-532
Anti-ENG Mouse Monoclonal Antibody (Biotin) [clone: 166707]
Antibodies
Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: 166707
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR