Anti-SIGLEC5 Mouse Monoclonal Antibody (Biotin) [clone: 194111]

Supplier: R & D Systems
BAM10722
220014-531EA 714.4 USD
220014-531
Anti-SIGLEC5 Mouse Monoclonal Antibody (Biotin) [clone: 194111]
Antibodies
Type: Primary
Antigen: SIGLEC5
Clonality: Monoclonal
Clone: 194111
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR