Anti-SPINT1 Mouse Monoclonal Antibody (Biotin) [clone: 169428]

Supplier: R & D Systems
BAM10482
220014-529EA 714.4 USD
220014-529
Anti-SPINT1 Mouse Monoclonal Antibody (Biotin) [clone: 169428]
Antibodies
Type: Primary
Antigen: SPINT1
Clonality: Monoclonal
Clone: 169428
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR