Anti-LY6G Rat Monoclonal Antibody (Biotin) [clone: RB6-8C5]

Supplier: R & D Systems
BAM1037
220014-528EA 728.86 USD
220014-528
Anti-LY6G Rat Monoclonal Antibody (Biotin) [clone: RB6-8C5]
Antibodies
Type: Primary
Antigen: LY6G
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR