Anti-FZD3 Rat Monoclonal Antibody (Biotin) [clone: 169310]

Supplier: R & D Systems
BAM1001
220014-527EA 552.43 USD
220014-527
Anti-FZD3 Rat Monoclonal Antibody (Biotin) [clone: 169310]
Antibodies
Type: Primary
Antigen Symbol: FZD3
Antigen Name: frizzled homolog 3 (Drosophila)
Antigen Synonyms: AU020229, D930050A07Rik, Fz3
Clonality: Monoclonal
Clone: 169310
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR