Anti-ANGPT2 Mouse Monoclonal Antibody (Biotin) [clone: 85834]

Supplier: R & D Systems
BAM0981
220014-526EA 769.35 USD
220014-526
Anti-ANGPT2 Mouse Monoclonal Antibody (Biotin) [clone: 85834]
Antibodies
Type: Primary
Antigen: ANGPT2
Clonality: Monoclonal
Clone: 85834
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR