Anti-Hamster IgG Mouse Monoclonal Antibody [clone: MAH1.12] (Biotin)

Supplier: R & D Systems
BAM011
220014-524EA 312.37 USD
220014-524
Anti-Hamster IgG Mouse Monoclonal Antibody [clone: MAH1.12] (Biotin)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: MAH1.12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR