Anti-AREG Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF989
220014-520EA 753.45 USD
220014-520
Anti-AREG Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: AREG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR