Anti-GMCSF1 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF987
220014-518EA 714.4 USD
220014-518
Anti-GMCSF1 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: GM-CSF
Antigen Name:
Antigen Synonyms:
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR