Anti-GAS6 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF986
220014-517EA 714.4 USD
220014-517
Anti-GAS6 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: GAS6
Antigen Name: growth arrest specific 6
Antigen Synonyms: Gas-6
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR