Anti-FGF19 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF969
220014-504EA 714.4 USD
220014-504
Anti-FGF19 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: FGF19
Antigen Name: fibroblast growth factor 19
Antigen Synonyms: -
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR