Anti-IFNG Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF961
220014-500EA 753.45 USD
220014-500
Anti-IFNG Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rhesus Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR