Anti-GDF3 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF958
220014-497EA 698.49 USD
220014-497
Anti-GDF3 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: GDF3
Antigen Name: growth differentiation factor 3
Antigen Synonyms: C78318, Gdf-3, Vgr-2, Vgr2, ecat9
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR