Anti-CTSB Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF953
220014-493EA 714.4 USD
220014-493
Anti-CTSB Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CTSB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR