Anti-SPARC Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF941
220014-483EA 698.49 USD
220014-483
Anti-SPARC Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: SPARC
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR