Anti-GDF15 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF940
220014-482EA 753.45 USD
220014-482
Anti-GDF15 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: GDF15
Antigen Name: growth differentiation factor 15
Antigen Synonyms: GDF-15, MIC-1, MIC1, NAG-1, PDF, PLAB, PTGFB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR