Anti-CTSZ Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF934
220014-477EA 698.49 USD
220014-477
Anti-CTSZ Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CTSZ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR